Anti-FKBP5 Rat Monoclonal Antibody [clone: 425314]

Supplier: R & D Systems
MAB4094
220020-744EA 584.25 USD
220020-744
Anti-FKBP5 Rat Monoclonal Antibody [clone: 425314]
Antibodies
Type: Primary
Antigen Symbol: FKBP5
Antigen Name: FK506 binding protein 5
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 425314
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR