Anti-REN Mouse Monoclonal Antibody [clone: 411525]

Supplier: R & D Systems
MAB4090
220020-742EA 633.42 USD
220020-742
Anti-REN Mouse Monoclonal Antibody [clone: 411525]
Antibodies
Type: Primary
Antigen: REN
Clonality: Monoclonal
Clone: 411525
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR