Anti-LAMP3 Mouse Monoclonal Antibody [clone: 530217]

Supplier: R & D Systems
MAB4087
220020-740EA 577.02 USD
220020-740
Anti-LAMP3 Mouse Monoclonal Antibody [clone: 530217]
Antibodies
Type: Primary
Antigen: LAMP3
Clonality: Monoclonal
Clone: 530217
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR