Anti-HAPLN4 Mouse Monoclonal Antibody [clone: 387915]

Supplier: R & D Systems
MAB4085
220020-739EA 577.02 USD
220020-739
Anti-HAPLN4 Mouse Monoclonal Antibody [clone: 387915]
Antibodies
Type: Primary
Antigen Symbol: HAPLN4
Antigen Name: hyaluronan and proteoglycan link protein 4
Antigen Synonyms: BRAL2
Clonality: Monoclonal
Clone: 387915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR