Anti-TFF2 Mouse Monoclonal Antibody [clone: 366508]

Supplier: R & D Systems
MAB4077
220020-738EA 600.15 USD
220020-738
Anti-TFF2 Mouse Monoclonal Antibody [clone: 366508]
Antibodies
Type: Primary
Antigen: TFF2
Clonality: Monoclonal
Clone: 366508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR