Anti-IL7 Rat Monoclonal Antibody [clone: 204626]

Supplier: R & D Systems
MAB407
220020-737EA 772.25 USD
220020-737
Anti-IL7 Rat Monoclonal Antibody [clone: 204626]
Antibodies
Type: Primary
Antigen: IL7
Clonality: Monoclonal
Clone: 204626
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR