Anti-FUT3 Mouse Monoclonal Antibody [clone: 425037]

Supplier: R & D Systems
MAB4068
220020-734EA 577.02 USD
220020-734
Anti-FUT3 Mouse Monoclonal Antibody [clone: 425037]
Antibodies
Type: Primary
Antigen Symbol: FUT3
Antigen Name: fucosyltransferase 3 (galactoside 3(4)-L-fucosyltransferase, Lewis blood group)
Antigen Synonyms: CD174, FT3B, FucT-III, LE, Les
Clonality: Monoclonal
Clone: 425037
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR