Anti-LAT2 Mouse Monoclonal Antibody [clone: 440005]

Supplier: R & D Systems
MAB4066
102885-680EA 561.11 USD
102885-680
Anti-LAT2 Mouse Monoclonal Antibody [clone: 440005]
Antibodies
Type: Primary
Antigen: LAT2
Clonality: Monoclonal
Clone: 440005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR