Anti-SERPINA6 Rat Monoclonal Antibody [clone: 433903]

Supplier: R & D Systems
MAB4065
220020-732EA 594.37 USD
220020-732
Anti-SERPINA6 Rat Monoclonal Antibody [clone: 433903]
Antibodies
Type: Primary
Antigen: SERPINA6
Clonality: Monoclonal
Clone: 433903
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR