Anti-ITGB4 Rat Monoclonal Antibody [clone: 308601]

Supplier: R & D Systems
MAB4054
220020-728EA 594.37 USD
220020-728
Anti-ITGB4 Rat Monoclonal Antibody [clone: 308601]
Antibodies
Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone: 308601
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR