Anti-NFAM1 Mouse Monoclonal Antibody [clone: 374301]

Supplier: R & D Systems
MAB4046
220020-726EA 577.02 USD
220020-726
Anti-NFAM1 Mouse Monoclonal Antibody [clone: 374301]
Antibodies
Type: Primary
Antigen Symbol: NFAM1
Antigen Name: NFAT activating protein with ITAM motif 1
Antigen Synonyms: CNAIP, bK126B4.4
Clonality: Monoclonal
Clone: 374301
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR