Anti-TFPI2 Rat Monoclonal Antibody [clone: 375717]

Supplier: R & D Systems
MAB4045
220020-725EA 561.11 USD
220020-725
Anti-TFPI2 Rat Monoclonal Antibody [clone: 375717]
Antibodies
Type: Primary
Antigen: TFPI2
Clonality: Monoclonal
Clone: 375717
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR