Anti-NAMPT Mouse Monoclonal Antibody [clone: 882104]

Supplier: R & D Systems
MAB40441
102885-678EA 577.02 USD
102885-678
Anti-NAMPT Mouse Monoclonal Antibody [clone: 882104]
Antibodies
Simple Western

Type: Primary
Antigen: NAMPT
Clonality: Monoclonal
Clone: 882104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR