Anti-NAMPT Rat Monoclonal Antibody [clone: 362616]

Supplier: R & D Systems
MAB4044
220020-724EA 594.37 USD
220020-724
Anti-NAMPT Rat Monoclonal Antibody [clone: 362616]
Antibodies
Type: Primary
Antigen: NAMPT
Clonality: Monoclonal
Clone: 362616
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR