Anti-TSC2 Mouse Monoclonal Antibody [clone: 614204]

Supplier: R & D Systems
MAB40401
102885-676EA 594.37 USD
102885-676
Anti-TSC2 Mouse Monoclonal Antibody [clone: 614204]
Antibodies
Type: Primary
Antigen: TSC2
Clonality: Monoclonal
Clone: 614204
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR