Anti-PLEKHA1 Rat Monoclonal Antibody [clone: 376409]

Supplier: R & D Systems
MAB4039
220020-722EA 561.11 USD
220020-722
Anti-PLEKHA1 Rat Monoclonal Antibody [clone: 376409]
Antibodies
Type: Primary
Antigen: PLEKHA1
Clonality: Monoclonal
Clone: 376409
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR