Anti-SMAD3 Rat Monoclonal Antibody [clone: 378611]

Supplier: R & D Systems
MAB4038
220020-721EA 600.15 USD
220020-721
Anti-SMAD3 Rat Monoclonal Antibody [clone: 378611]
Antibodies
Type: Primary
Antigen: SMAD3
Clonality: Monoclonal
Clone: 378611
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR