Anti-SMAD2 Mouse Monoclonal Antibody [clone: 376520]

Supplier: R & D Systems
MAB4037
220020-720EA 577.02 USD
220020-720
Anti-SMAD2 Mouse Monoclonal Antibody [clone: 376520]
Antibodies
Type: Primary
Antigen: SMAD2
Clonality: Monoclonal
Clone: 376520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR