Anti-PROS1 Mouse Monoclonal Antibody [clone: 391609]

Supplier: R & D Systems
MAB4036
220020-719EA 577.02 USD
220020-719
Anti-PROS1 Mouse Monoclonal Antibody [clone: 391609]
Antibodies
Type: Primary
Antigen: PROS1
Clonality: Monoclonal
Clone: 391609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR