Anti-C1QA Rat Monoclonal Antibody [clone: 394107]

Supplier: R & D Systems
MAB4035
220020-718EA 577.02 USD
220020-718
Anti-C1QA Rat Monoclonal Antibody [clone: 394107]
Antibodies
Type: Primary
Antigen: C1QA
Clonality: Monoclonal
Clone: 394107
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR