Anti-KLK12 Mouse Monoclonal Antibody [clone: 364926]

Supplier: R & D Systems
MAB4034
220020-716EA 561.11 USD
220020-716
Anti-KLK12 Mouse Monoclonal Antibody [clone: 364926]
Antibodies
Type: Primary
Antigen: KLK12
Clonality: Monoclonal
Clone: 364926
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR