Anti-HSPH1 Mouse Monoclonal Antibody [clone: 531104]

Supplier: R & D Systems
MAB4029
220020-714EA 577.02 USD
220020-714
Anti-HSPH1 Mouse Monoclonal Antibody [clone: 531104]
Antibodies
Type: Primary
Antigen: HSPH1
Clonality: Monoclonal
Clone: 531104
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR