Anti-MYOCD Mouse Monoclonal Antibody [clone: 355521]

Supplier: R & D Systems
MAB4028
220020-713EA 623.29 USD
220020-713
Anti-MYOCD Mouse Monoclonal Antibody [clone: 355521]
Antibodies
Type: Primary
Antigen: MYOCD
Clonality: Monoclonal
Clone: 355521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR