Anti-MYF5 Mouse Monoclonal Antibody [clone: 593128]

Supplier: R & D Systems
MAB4027
102885-674EA 594.37 USD
102885-674
Anti-MYF5 Mouse Monoclonal Antibody [clone: 593128]
Antibodies
Type: Primary
Antigen: MYF5
Clonality: Monoclonal
Clone: 593128
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR