Anti-IL2 Rat Monoclonal Antibody [clone: 142206]

Supplier: R & D Systems
MAB402
220020-712EA 772.25 USD
220020-712
Anti-IL2 Rat Monoclonal Antibody [clone: 142206]
Antibodies
Type: Primary
Antigen: IL2
Clonality: Monoclonal
Clone: 142206
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR