Anti-IRF3 Mouse Monoclonal Antibody [clone: 482205]

Supplier: R & D Systems
MAB4019
220020-711EA 594.37 USD
220020-711
Anti-IRF3 Mouse Monoclonal Antibody [clone: 482205]
Antibodies
Type: Primary
Antigen: IRF3
Clonality: Monoclonal
Clone: 482205
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR