Anti-BCAN Mouse Monoclonal Antibody [clone: 450616]

Supplier: R & D Systems
MAB40091
220020-707EA 577.02 USD
220020-707
Anti-BCAN Mouse Monoclonal Antibody [clone: 450616]
Antibodies
Type: Primary
Antigen: BCAN
Clonality: Monoclonal
Clone: 450616
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR