Anti-IL1A Hamster Monoclonal Antibody [clone: ALF161]

Supplier: R & D Systems
MAB4001
220020-705EA 763.57 USD
220020-705
Anti-IL1A Hamster Monoclonal Antibody [clone: ALF161]
Antibodies
Type: Primary
Antigen: IL1A
Clonality: Monoclonal
Clone: ALF161
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR