Anti-EIF2S1 Rat Monoclonal Antibody [clone: 849159]

Supplier: R & D Systems
MAB39971
102885-670EA 594.37 USD
102885-670
Anti-EIF2S1 Rat Monoclonal Antibody [clone: 849159]
Antibodies
Type: Primary
Antigen: EIF2S1
Clonality: Monoclonal
Clone: 849159
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR