Anti-EIF2S1 Rat Monoclonal Antibody [clone: 430516]

Supplier: R & D Systems
MAB3997
220020-703EA 577.02 USD
220020-703
Anti-EIF2S1 Rat Monoclonal Antibody [clone: 430516]
Antibodies
Type: Primary
Antigen: EIF2S1
Clonality: Monoclonal
Clone: 430516
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR