Anti-BCL9 Mouse Monoclonal Antibody [clone: 417514]

Supplier: R & D Systems
MAB3996
220020-702EA 594.37 USD
220020-702
Anti-BCL9 Mouse Monoclonal Antibody [clone: 417514]
Antibodies
Type: Primary
Antigen: BCL9
Clonality: Monoclonal
Clone: 417514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR