Anti-PARK7 Rat Monoclonal Antibody [clone: 421015]

Supplier: R & D Systems
MAB3995
220020-701EA 577.02 USD
220020-701
Anti-PARK7 Rat Monoclonal Antibody [clone: 421015]
Antibodies
Type: Primary
Antigen: PARK7
Clonality: Monoclonal
Clone: 421015
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR