Anti-IRS1 Mouse Monoclonal Antibody [clone: 738710]

Supplier: R & D Systems
MAB39783
102885-666EA 577.02 USD
102885-666
Anti-IRS1 Mouse Monoclonal Antibody [clone: 738710]
Antibodies
Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: 738710
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR