Anti-IRS1 Mouse Monoclonal Antibody [clone: 614115]

Supplier: R & D Systems
MAB39782
102885-664EA 577.02 USD
102885-664
Anti-IRS1 Mouse Monoclonal Antibody [clone: 614115]
Antibodies
Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: 614115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR