Anti-IRS1 Mouse Monoclonal Antibody [clone: 395601]

Supplier: R & D Systems
MAB39781
220020-698EA 584.25 USD
220020-698
Anti-IRS1 Mouse Monoclonal Antibody [clone: 395601]
Antibodies
Type: Primary
Antigen: IRS1
Clonality: Monoclonal
Clone: 395601
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR