Anti-NPR1 Rat Monoclonal Antibody [clone: 488620]

Supplier: R & D Systems
MAB3974
220020-697EA 577.02 USD
220020-697
Anti-NPR1 Rat Monoclonal Antibody [clone: 488620]
Antibodies
Type: Primary
Antigen: NPR1
Clonality: Monoclonal
Clone: 488620
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR