Anti-DLL1 Mouse Monoclonal Antibody [clone: 409005]

Supplier: R & D Systems
MAB3970
220020-695EA 584.25 USD
220020-695
Anti-DLL1 Mouse Monoclonal Antibody [clone: 409005]
Antibodies
Type: Primary
Antigen: DLL1
Clonality: Monoclonal
Clone: 409005
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR