Anti-ADGRB3 Mouse Monoclonal Antibody [clone: 49611]

Supplier: R & D Systems
MAB3965
220020-692EA 566.89 USD
220020-692
Anti-ADGRB3 Mouse Monoclonal Antibody [clone: 49611]
Antibodies
Type: Primary
Antigen: ADGRB3
Clonality: Monoclonal
Clone: 49611
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR