Anti-S1PR5 Mouse Monoclonal Antibody [clone: 282503]

Supplier: R & D Systems
MAB3964
220020-691EA 577.02 USD
220020-691
Anti-S1PR5 Mouse Monoclonal Antibody [clone: 282503]
Antibodies
Type: Primary
Antigen: S1PR5
Clonality: Monoclonal
Clone: 282503
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR