Anti-B3GNT2 Mouse Monoclonal Antibody [clone: 485327]

Supplier: R & D Systems
MAB3960
220020-690EA 566.89 USD
220020-690
Anti-B3GNT2 Mouse Monoclonal Antibody [clone: 485327]
Antibodies
Type: Primary
Antigen: B3GNT2
Clonality: Monoclonal
Clone: 485327
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR