Anti-AOC3 Mouse Monoclonal Antibody [clone: 393112]

Supplier: R & D Systems
MAB3957
220020-688EA 594.37 USD
220020-688
Anti-AOC3 Mouse Monoclonal Antibody [clone: 393112]
Antibodies
Type: Primary
Antigen: AOC3
Clonality: Monoclonal
Clone: 393112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR