Anti-FKBP3 Rat Monoclonal Antibody [clone: 423325]

Supplier: R & D Systems
MAB3955
220020-687EA 577.02 USD
220020-687
Anti-FKBP3 Rat Monoclonal Antibody [clone: 423325]
Antibodies
Type: Primary
Antigen Symbol: FKBP3
Antigen Name: FK506 binding protein 3
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 423325
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR