Anti-EFNB3 Mouse Monoclonal Antibody [clone: 88838]

Supplier: R & D Systems
MAB395
220020-686EA 763.57 USD
220020-686
Anti-EFNB3 Mouse Monoclonal Antibody [clone: 88838]
Antibodies
Type: Primary
Antigen: EFNB3
Clonality: Monoclonal
Clone: 88838
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR