Anti-F2RL1 Mouse Monoclonal Antibody [clone: 344222]

Supplier: R & D Systems
MAB3949
220020-685EA 655.11 USD
220020-685
Anti-F2RL1 Mouse Monoclonal Antibody [clone: 344222]
Antibodies
Type: Primary
Antigen: F2RL1
Clonality: Monoclonal
Clone: 344222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR