Anti-ACP5 Mouse Monoclonal Antibody [clone: 41763]

Supplier: R & D Systems
MAB3948
220020-684EA 577.02 USD
220020-684
Anti-ACP5 Mouse Monoclonal Antibody [clone: 41763]
Antibodies
Type: Primary
Antigen: ACP5
Clonality: Monoclonal
Clone: 41763
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR