Anti-ST14 Mouse Monoclonal Antibody [clone: 416802]

Supplier: R & D Systems
MAB3946
220020-680EA 600.15 USD
220020-680
Anti-ST14 Mouse Monoclonal Antibody [clone: 416802]
Antibodies
Type: Primary
Antigen: ST14
Clonality: Monoclonal
Clone: 416802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR