Anti-FCAR Mouse Monoclonal Antibody [clone: 488032]

Supplier: R & D Systems
MAB3939
220020-678EA 600.15 USD
220020-678
Anti-FCAR Mouse Monoclonal Antibody [clone: 488032]
Antibodies
Type: Primary
Antigen Symbol: FCAR
Antigen Name: Fc fragment of IgA receptor
Antigen Synonyms: CD89, CTB-61M7.2, FcalphaRI
Clonality: Monoclonal
Clone: 488032
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR