Anti-CRHR1 Mouse Monoclonal Antibody [clone: 343919]

Supplier: R & D Systems
MAB3930
220020-676EA 594.37 USD
220020-676
Anti-CRHR1 Mouse Monoclonal Antibody [clone: 343919]
Antibodies
Type: Primary
Antigen: CRHR1
Clonality: Monoclonal
Clone: 343919
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR