Anti-PPBP Mouse Monoclonal Antibody [clone: 59418]

Supplier: R & D Systems
MAB393
220020-675EA 779.48 USD
220020-675
Anti-PPBP Mouse Monoclonal Antibody [clone: 59418]
Antibodies
Simple Western

Type: Primary
Antigen: PPBP
Clonality: Monoclonal
Clone: 59418
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR