Anti-ANXA2 Mouse Monoclonal Antibody [clone: 666316]

Supplier: R & D Systems
MAB3928
102885-662EA 600.15 USD
102885-662
Anti-ANXA2 Mouse Monoclonal Antibody [clone: 666316]
Antibodies
Simple Western

Type: Primary
Antigen: ANXA2
Clonality: Monoclonal
Clone: 666316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR